Description
- A unique one size earplugs which fits most ear canals comfortably and provides a high level of attenuation
- Reusable plug is comfortable, hygienic and economical as it can be washed
- Triple-flange design preformed to fit the ear
Specifications
Specifications
Color | Yellow, Blue |
Pairs per Package | 50 Pairs/Box |
Noise Reduction Rating (NRR) | 35 dB |
Corded | Corded |
Dielectric | No |
Material | Elastomeric Polymer |
Product Content Correction
Product Title
3M™ E-A-R™ Ultrafit™ X Earplugs >
Spot an opportunity for improvement?Share a Content Correction