Learn More
3M™ E-A-R™ UltraTech Earplugs
3M™ E-A-R™ UltraTech Earplugs greatly improve the ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively.
Brand: 3M™ 7000103799
Additional Details : Weight : 0.00810kg
Description
- Improve wearer's ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively
- Earplug are ideal for musicians who require a flat attenuation protection
- Conical shape plugs help provide effective seal for reassured protection
- Effective protection against moderate levels of harmful noise SNR 21 dB
Specifications
Black, White | |
21 dB | |
No |
1 Pair/Box | |
Uncorded |