Learn More
3M™ E-A-Rfit™ Probed Test Ear Plugs
Product Code. 15799777
Click to view available options
Model:
393-2001-50
393-2002-50
393-2004-50
393-2007-50
393-2008-50
393-2010-50
Size:
Small
Universal
Product Type:
Classic™ Small
Express™
FX™
Model 1100
Push-Ins™
UltraFit™
Test ear plugs are modified with a probe tube for use during ear plug fit testing with the 3M™ E-A-Rfit™ Validation System
  • Science-based, quantitative testing gives accurate reports of hearing protection
  • Fast, clear, accurate results are obtained in just seconds
  • Test ear plugs are modified with a probe tube for use during ear plug fit testing with the 3M™ E-A-Rfit™ Validation System
  • 50 pair per case
  • 3M PSD products are for occupational use only
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
For Use With (Application) Health & Safety Fit Testing
High Visibility No
Model 393-2008-50
Packaging Type Poly bag
Pairs per Package 50 Pairs/Cs
Plug Material Polyurethane
Plug Color Multi-color
Corded Uncorded
Plug Shape Pod
Product Type Express™
Metal Detectable No