missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences pASG-IBA E. coli Expression Vector with No Secretion

Regulate the expression of proteins with a vector that carries the inducible tetracycline promoter/operator. IBA Lifesciences GmbH E. coli Expression Vector with No Secretion allows for the expression of proteins in E.coli.

Brand:  IBA Lifesciences 5-4005-001

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15683728

  • £229.00 / 5µg
Estimated Shipment: 25-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • The pASG-IBA vectors are designed for expression of target proteins without affinity tag in E. coli.
  • The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins.
  • The vectors can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli.
  • Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity.
Please note that cloning into pASG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pASG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.
Specifications

Specifications

Inducible with anhydrotetracycline
Tet
Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Strep-tag II
250 ng/μL
5 μg
Vector for recombinant expression in E. coli
Ampicillin
Vector
E.coli
Direct Cloning
Forward
pASG-IBA5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers