Learn More
Invitrogen™ E-Gel™ 1 Kb Plus Express DNA Ladder

Ready-to use DNA Ladder for use with routine throughput precast E-Gel™ agarose gels.
Brand: Invitrogen™ 10488091
198.64 GBP valid until 2025-12-31
Use promo code "24358" to get your promotional price.
Includes: DNA Ladder and 1 vial 1X E-Gel™ Sample Loading Buffer
Description
Invitrogen E-Gel 1 Kb Plus Express DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 5,000 bp. E-Gel 1 Kb Plus Express DNA Ladder consists of nine individual chromatography-purified DNA fragments with reference bands at 1,500 and 500 bp for easy orientation.
E-Gel 1 Kb Plus Express DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.
Highlights of E-Gel 1 Kb Plus Express DNA Ladder:
- Performance—optimized for use on E-Gel CloneWell ll agarose gels
- Sharp, clear bands—chromatography-purified fragments for consistent and reliable results, stable at room temperature up to six months
- Ready to use—premixed with loading buffer
- Convenient—provided with 1X E-Gel Sample Loading Buffer
- Precise—an exact amount of DNA in each band

Specifications
• 2 x 1.25 mL E-Gel 1 Kb Plus Express DNA Ladder • 1.25 mL 1X E-Gel Sample Loading Buffer Store at room temperature or at 4°C for up to 6 months. For longer periods store at -20°C. |
|
Agarose | |
Yes | |
100 to 5000 bp | |
CloneWell™ II E-Gels, Single Comb E-Gels | |
100 applications | |
Individual chromatography-purified DNA fragments |
0.04 μg/μL | |
DNA Ladder | |
1.25 mL | |
2 x 1.25 mL | |
Sustainable packaging | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.