Learn More
Invitrogen™ E-Gel™ 48 Agarose Gels
Invitrogen E-Gel 48 high-throughput precast agarose gels with or without SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis.
Brand: Invitrogen™ G820844
675.92 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 2.80000kg
Description
Invitrogen E-Gel 48 high-throughput precast agarose gels with or without SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.
Each gel contains 48 sample wells and 4 marker wells in 1%, 2%, or 4% high-resolution agarose with a 3.2-cm run length. The various agarose percentages are used to separate DNA fragments of different sizes:
- 1% agarose gels: DNA fragments between 400 bp and 10 kb
- 2% agarose gels: DNA fragments between 50 bp and 3 kb
- 4% agarose gels: DNA fragments between 10 bp and 500 bp
Each cassette is individually labeled with an EAN39 bar code that is recognized by most commercially available robotic bar code readers.
Features of E-Gel gels:
- High-throughput analysis—contains 48 sample lanes and 4 marker lanes per gel
- Automation compatible—suitable for use with multi-channel pipettors and commonly used 8- and 12-pin liquid handling robots
- Scalable—resolve up to 192 samples in one run by combining up to four E-Base Mother and E-Base Daughter device units
- Less hazardous choice—contains SYBR Safe DNA stain, a less hazardous alternative to ethidium bromide
- High performance—detect 3 ng of sample DNA with separation time as little as ∼20 minutes
Less hazardous option
Choose gels with SYBR Safe DNA stain to minimize exposure to hazardous and mutagenic factors such as ethidium bromide and UV. Precast E-Gel agarose gels with SYBR Safe stain, combined with a blue-light trans-illumination, offer better sample integrity and a less hazardous work environment. Unlike ethidium bromide, the DNA stain most commonly incorporated into agarose gels, SYBR Safe DNA Gel Stain is a non-toxic, non-mutagenic DNA stain that is less hazardous for the user, less hazardous for the environment, and can save institutional overhead costs for disposal.
Fast and convenient analysis
E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng sample DNA per band and a separation time three times faster than that of conventional pour-your-own gels. Each E-Gel agarose gel contains agarose, electrodes, SYBR Safe DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.
Running and imaging devices
High-throughput E-Gel agarose gels require the Invitrogen E-Base Electrophoresis Device for gel running.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The high-throughput E-Gel 96 and 48 agarose gels with SYBR Safe stain are available in 1%, 2%, and 4% gel percentages.
Specifications
E-Gel™ Ultra Low Range DNA Ladder | |
4% | |
Agarose Gel | |
SYBR™ Safe | |
Less hazardous | |
Room Temperature |
• Each box contains 8 gels Store strictly at room temperature. Do not freeze. Avoid cold spots near freezers or other uncontrolled environments. |
|
E-Gel | |
10 to 500 bp | |
48-well | |
4 x 8 Gels | |
Double Comb |
For Research Use Only. Not for use in diagnostic procedures.