missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 2%

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 2%, provides everything you need to quickly start using precast 1% E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the system for a streamlined gel electrophoresis and gel imaging workflow.

Brand:  Invitrogen™ G8372ST

2110.10 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


Product Code. 17135989

  • £3165.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

The kit includes:

  • Ten E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
  • E-Gel™ Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel™ Low Range Quantitative DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 12373031)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel Low Range Quantitative DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 12373031)

Store at room temperature.
Mini
Electrophoresis System Starter Kit
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.