Learn More
Invitrogen™ E-PAGE™ Midi Protein Gels, 3.7 mm
E-PAGE™ Gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable cassette.
Brand: Invitrogen™ EP04808
355.20 GBP valid until 2024-12-31
Use promo code "21959" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Includes: Eight gels, E-PAGE™ loading buffer 1, one reusable E-PAGE™ Blotting pad, and a Butterfly Opener.
Description
E-PAGE Gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable cassette. Protein separation on an E-PAGE Gel requires an EBase Integrated Device, a compact and automated platform designed for medium- and high-throughput electrophoresis.
Each E-PAGE 8% 48-well gel contains 48 sample lanes and 4 marker lanes with a 3.2-cm separation length. E-PAGE 48-well gels can be loaded with a 12- or 8-channel pipettor or 4- or 8-tip robotic system (Figure 1).
Each E-PAGE 6% 96-well gel contains 96 sample lanes and 8 marker lanes with a 1.6-cm separation length. The well openings of the cassette are compatible with the standard 96-well plate format and can be conveniently loaded with a multichannel pipettor or 8-, 12-, or 96-tip liquid-handling robotic device (Figure 2).
Both E-PAGE 48 and 96 cassettes can be opened to remove the gel for downstream staining or blotting (Figures 3 and 4). In addition, each cassette is labeled with an individual barcode to facilitate identification of the gel using commercial barcode readers.
More about the E-PAGE precast gel system
The Invitrogen E-PAGE precast gel system is designed for fast, mess-free medium- and high-throughput protein electrophoresis. It consists of the E-PAGE precast gels, E-Base Integrated devices for running the gels, and free E-Editor 2.0 software for image analysis.
The all-in-one E-Gel Power Snap Plus Electrophoresis System is designed for fast and convenient high-throughput gel electrophoresis. With dry precast E-PAGE gel technology, you can run protein samples in as little as 14 minutes. The device arrives with pre-programmed protocols for both E-PAGE gel types.
Order Info
Shipping Condition: Room Temperature
Specifications
E-BASE | |
Midi | |
E-PAGE | |
20 to 220 kDa | |
48-well | |
E-PAGE™ | |
Room Temperature | |
8 cm |
8% | |
3.7 mm | |
Up to 15 μL | |
Denaturing | |
Molecular Weight | |
8 Gels/Box | |
Store at room temperature, do not allow the temperature to drop below 4°C or rise above 40°C | |
13 cm |