missing translation for 'onlineSavingsMsg'
Learn More

E2F7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen E2F7
Concentration 0.38 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16802965
View Documents
Proteintech
24489-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16892955
View Documents
Proteintech
24489-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression.
Specifications

Specifications

E2F7
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
Q96AV8
144455
E2F7 Fusion Protein Ag21599
Primary
-20°C
E2F7
0.38 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
E2F transcription factor 7, E2F7
E2F7
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications