Learn More
Abeba™ Easy 37622 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
£56.68
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | White |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15631098
|
Abeba™
37622/35 |
35 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15641098
|
Abeba™
37622/36 |
36 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15651098
|
Abeba™
37622/37 |
37 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15661098
|
Abeba™
37622/38 |
38 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15671098
|
Abeba™
37622/39 |
39 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15681098
|
Abeba™
37622/40 |
40 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15691098
|
Abeba™
37622/41 |
41 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15601108
|
Abeba™
37622/42 |
42 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15611108
|
Abeba™
37622/43 |
43 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15621108
|
Abeba™
37622/44 |
44 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15631108
|
Abeba™
37622/45 |
45 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15641108
|
Abeba™
37622/46 |
46 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15651108
|
Abeba™
37622/47 |
47 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15661108
|
Abeba™
37622/48 |
48 |
£56.68
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white/blue
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3573)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Polyurethane |
White | |
Slip-resistant | |
37622 |