Learn More
Abeba™ Easy 7622 Shoes
Includes TPU Sole with high slip resistance. Upper leather and microfiber construction.
Brand:  Abeba™ 7622/45
Additional Details : Weight : 0.97000kg
 View more versions of this product
Product Code. 15672888
  • £54.51 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white/blue
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
7622
White
45
Polyurethane