missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EASY-Spray™ HPLC Columns

Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.

Brand:  Thermo Scientific™ ES906

 View more versions of this product

Product Code. 16622037

  • £1470.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.

The unique EASY-Spray design provides uncompromised performance and ease-of-use in nano and capillary LC-MS proteomics applications. Features include the following:

  • Precision positioned glass emitter
  • Integrated precision designed union
  • Integrated temperature control
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column, EASY-Spray
150 mm
2 μm
100 Å
60°C
C18
Spherical, Fully Porous, Ultrapure Silica
Reversed Phase
Reversed Phase
150 μm
14500 psi (1000 bar)
2 to 8
450 m²/g
L1
Easy Spray
15%
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.