missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EASY-Spray™ HPLC Columns

Product Code. 16632037 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
150 mm
250 mm
500 mm
750 mm
Diameter (Metric):
50 μm
75 μm
150 μm
200 μm
Particle Size:
2 μm
2.6 μm
3 μm
4 μm
5 μm
Monolith
Packing Material:
Monolith Polymer
PSDVB (Polystyrene Divinylbenzene)
Spherical, Fully Porous, Ultrapure Silica
Spherical, Solid Core, Ultrapure Silica
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16632037

Brand: Thermo Scientific™ ES911

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.

Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.

The unique EASY-Spray design provides uncompromised performance and ease-of-use in nano and capillary LC-MS proteomics applications. Features include the following:

  • Precision positioned glass emitter
  • Integrated precision designed union
  • Integrated temperature control
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column, EASY-Spray
Diameter (Metric) 75 μm
Length (Metric) 150 mm
Max. Pressure 11600 psi (800 bar)
Particle Size 2.6 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 80 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Product Line Easy Spray
Packing Material Spherical, Solid Core, Ultrapure Silica
Carbon Load 2%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.