Learn More
Thermo Scientific™ Easy-Titer™ Rabbit IgG Assay Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Easy-Titer Antibody Assay Kits enable accurate determination of antibody concentration in serum and culture supernatant, and include kits specific for mouse IgG, rabbit IgG, human IgG (H+L), human IgG (Fc), or human IgM.
Highlights:
Kits for five popular targets – choose a kit specific for a particular species and class of immunoglobulin; no cross-reaction with other species and classes of the target antibody
Antibody-based specificity – measure concentration of target antibody in a sample, not just total protein; no need to purify antibody to assess its concentration
Faster and easier than ELISA – three-component, homogenous assay; 10 minutes total incubation time
No special equipment needed – uses standard vortex mixer, pipetter, 96-well microplate, plate shaker and reader (measure absorbance at 340nm or 400nm)
Sensitive – assay range (standard curve) 8 to 500ng/mL; requires starting sample of at least 300ng/mL
Reproducible – coefficient of variation less than 5%; error depends on dilution and pipetting technique
Antibody standards sold separately – use any antibody standard with proper target identity and known concentration (greater than 10μg/mL)
Easy-Titer Antibody Assays are faster and easier than ELISA:
1. Prepare standards (5 to 500ng/mL) by diluting purified antibody in Kit Dilution Buffer
2. Prepare samples by diluting at least 20-fold in Dilution Buffer to within assay range (8 to 500ng/mL).
3. Vortex vial of microsphere beads to create homogeneous suspension.
4. Pipette 20μL of bead suspension and 20μL of each sample and standard into 96-well microplate wells.
5. Incubate microplate for 5 minutes with vigorous mixing.
6. Add 100μL of Kit Blocking Reagent.
7. Incubate microplate for 5 minutes with vigorous mixing.
8. Measure absorbance on standard plate reader (340nm or 405nm).
9. Plot standard curve and interpolate samples to determine concentration.
Includes:
Each kit contains target-specific IgG sensitized microspheres, dilution buffer and blocking buffer
Specifications
Specifications
| Description | Easy-Titer Rabbit IgG Assay Kit |
| Content And Storage | Sufficient For: 96 total tests (including 9 standards per use) • Goat Anti-Rabbit IgG Sensitized Beads, 2mL • Easy-Titer Dilution Buffer, 30 mL • Easy-Titer Blocking Buffer, 15 mL Store at 4°C (do not freeze). |
| For Use With (Application) | Solution-based Detection, Absorbance |
| For Use With (Equipment) | Spectrophotometer, Microplate Reader |
| Quantity | 96 Tests |
| Assay | Protein Quantitation Assay, Isotyping Assay, Immunoassay |
| Detection Method | Colorimetric |
| Product Line | Easy-Titer |
| Target Specificity | Target-specific |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.