Learn More
Berkshire™ EasyClean™ 360 Telescoping aluminum Handle
BIOPHARMA
The EasyClean™ 360 Isolator Cleaning System includes a light-weight, maneuverable telescoping aluminum handle
Brand:  Berkshire™ HANDLEFMTELE180
Product Code. 13230644
  • £628.00 / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
95 to 180 cm
ISO Class 3 to 8, Cleanroom Class 1 to 100,000, EU Grade A/B/C/D
Aluminum