missing translation for 'onlineSavingsMsg'
Learn More

Contec™ DualClean™ Softwall Strip Cleaner System
GREENER_CHOICEBIOPHARMA

Designed to thoroughly and economically clean vinyl strip curtains with ease. DualClean™ Softwall Strip Cleaner System is a unique system that enables one operator to quickly clean both sides of a 4 to 12 in. plastic strip curtains in a single pass.

Brand:  Contec™ 2778

295.99 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15606986

  • £364.00 / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

DualClean System:

The system includes stainless steel frame with two curved-surface arms, fabric covers and a QuickConnect™ handle. QuickConnect handles are interchangeable. The recommended handle for the DualClean system is cat. no. 19168806, extendable fiberglass handle. All parts are sold separately.
  • Easily and uniformly cleans both sides of the length of the strip
  • Two fabric covers available to meet cleaning needs:
    • Microfiber terrycloth removes and traps dust and other visible contaminants
    • Quiltec™ knit polyester is ideally suited to critical environments
  • Frame attaches to a wide variety of Contec handles
  • Electropolished frame is autoclavable and resistant to most chemicals and disinfectants
Specifications

Specifications

36.5 x 17 cm (14.5 x 6.8 in.)
White
14.5 in.
36.8 cm
6.8 in.
Enables one operator to easily and quickly clean both sides of the plastic strip curtains in a single pass
Stainless Steel (304)
Dual Clean Frame
2778
Non-sterile
17 cm
Reusable
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers