missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EasyPep™ Magnetic MS Sample Preparation Kits and Proteomics Magnetic Clean-up Beads

Product Code. 18416458 Shop All Thermo Scientific Products
Click to view available options
Quantity:
20 Reactions
5 mL
96 Reactions
Product Type:
Magnetic Clean-up Beads
Magnetic MS Sample Prep Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18416458

Brand: Thermo Scientific™ A57868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EasyPep Magnetic MS Sample Preparation Kits and Proteomics Magnetic Clean-up Beads contain MS-compatible magnetic beads specifically designed to tolerate harsh MS sample prep conditions to improve sample quality and reproducibility for both manual and automated sample preparation methods.

Proteomic sample preparation for MS analysis is complex, resulting in challenges with reproducibility and peptide quality. For MS sample preparation methods that utilize magnetic beads, a major challenge is the compatibility of the beads with the harsh conditions required for sample processing. When exposed to extreme pH or solvents, traditional magnetic beads often clump or dissolve, resulting in increased variability.

Pierce Proteomics Magnetic Clean-up Beads have been designed for compatibility with common solvents (e.g. acetonitrile, ethanol, isopropyl alcohol, and acetone) and wide range of pH (e.g. pH 2-13) to enable fast, efficient, and reproducible removal of a wide variety of detergents, salts, and other mass spectrometry (MS)-incompatible contaminants from protein and peptide samples. The beads can be successfully used to clean up proteins derived from different sample types (e.g. purified proteins, cell lysates, tissue extracts, and biological fluids) and protein input amounts (e.g. 1 µg to 1 mg) using manual magnetic processing or automated workflows. 

EasyPep Magnetic MS Sample Prep Kits integrate optimized EasyPep sample preparation chemistry with the Proteomics Magnetic Clean-up Beads to support manual and automated magnetic bead sample processing, including the King Fisher Instrument Series. The complete kits contain pre-formulated buffers, MS-grade enzyme mix, magnetic beads for peptide clean-up, and an optimized protocol to generate MS-ready peptide samples in less than 3 hours. The kits are suitable for processing protein samples from 15–100 µg with a high yield of detergent-free peptides ready for direct LC-MS analysis without the need to SpeedVac dry and reconstitute before injection.  In addition, the peptide samples are compatible with isobaric tag (e.g., TMT/TMTpro Reagents) labeling and high pH reversed-phase fractionation.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Magnetic Clean-up Beads
Product Line Pierce
Starting Material Protein Samples
Detection Method Mass Spectrometry
Format Bottle
Quantity 5 mL
Form Liquid

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.