missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EasyPep™ MS Sample Prep Kits

Product Code. 17185772 Shop All Thermo Scientific Products
Click to view available options
Description:
Lysis Buffer
MS Sample Prep Kit
Maxi Sample Prep Kit
Micro MS Sample Prep Kit
Mini MS Sample Prep Kit
Peptide Clean-up Plate
Unit Size:
122g
20 tests
8 reactions
96 reactions
This item is not returnable. View return policy

Product Code. 17185772

Brand: Thermo Scientific™ A45734

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Generate high quality proteomic samples for MS analysis in less than four hours using optimized EasyPep MS Sample Prep kits.

EasyPep Mini MS Sample Prep Kits enable efficient and reproducible processing of cultured mammalian cells and tissues for proteomic MS analysis. The kit contains pre-formulated buffers, MS-grade enzyme mix, peptide clean-up columns, and an optimized protocol to generate MS-compatible peptide samples in less than three hours. Kits are optimized to process protein samples from 10–100 μg with a high yield of MS-ready peptides.

Complete—includes pre-formulated reagents for lysis through digestion, peptide clean-up columns, and an optimized protocol for up to 20 samples
Optimized—streamlined protocol and reagents minimize the number of steps and time time needed to process samples
Flexible—reagents and protocol have been verified using cells, plasma, and tissue samples for 10 to 100 μg samples
Time-saving—sample processing time reduced from more than one day to less than three hours
Compatible—final preparation is ready for direct MS analysis and other downstream applications, including TMT labeling

The sample preparation of peptides for MS analysis is complex, with numerous steps and varied protocols, resulting in sample variability in both quality and quantity. This kit has been designed to improve reproducibility while saving hands-on and processing time. The number of steps and processing time have been reduced through the addition of Universal Nuclease to reduce viscosity from nucleic acids (replacing sonication), a rapid 'one pot' reduction/alkylation solution for cysteine modification (carbamidomethylation, + 57.02), and a trypsin/Lys-C protease mix for more complete digestion. In addition, the kit includes Peptide Clean-up columns and buffers to prepare detergent-free peptide samples for MS analysis. The optimized reagents and protocol produce high-quality peptides that are compatible with TMT labeling and other downstream applications, or are ready for MS analysis.

TRUSTED_SUSTAINABILITY

Specifications

Description Maxi Sample Prep Kit
For Use With (Equipment) Mass Spectrometer
Product Type Sample Preparation Kit
Product Line EasyPep
Content And Storage
  • Lysis Solution, 25 mL, store at 4°C
  • Universal Nuclease, 25 kU, store at -20°C
  • Reduction Solution, 7 mL, store at 4°C
  • Alkylation Solution, 7 mL, store at 4°C
  • Enzyme Reconstititution solution, 7 mL, store at 4°C
  • Pierce™ Trypsin/Lys-C Protease Mix, MS Grade, 8 x 100 μg, store at -20°C
  • Digestion Stop Solution, 7 mL, store at 4°C
  • Peptide Clean-Up Columns, 8 Ea., store at 4°C
  • Wash Solution A, 40 mL, store at 4°C
  • Wash Solution B, 3 x 27 mL, store at 4°C
  • Elution Solution, 2 x 20 mL, store at 4°C
  • Low Protein Binding Collection Tubes, 2 mL, 40 Ea., store at 4°C

Label or Dye Unlabeled
Shipping Condition Wet Ice
Starting Material Protein Samples
Final Product Type Peptides
Detection Method Mass Spectrometry
Workflow Step Protein Digestion
Quantity 8 Reactions
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.