missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Easyplate Petri Dish Rack

Product Code. 10358469 Shop All SP Scienceware Products
Click to view available options
Color:
Assorted
Blue
Natural
Height (Metric):
24 cm
37 cm
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 10358469

Brand: Bel-Art™ Scienceware™ F189820409

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Increase workspace and efficiently transport large quantity of Petri dishes.

  • Silicone holding strips grip each Petri dish; easily insert and remove groups of dishes without disturbing upper or lower dishes
  • Side labels provide space for critical sample identification
  • Lightweight, chemical resistant polycarbonate
  • Autoclavable at 121°C (250°F)

Specifications

Product Type Petri Dish Rack
Autoclavable Autoclavable
Color Assorted
Holds Petri Dishes (100 mm)
Material Polycarbonate
Positions 48
For Use With (Equipment) Petri Dish
Quantity 1 Each
Height (Metric) 24 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.