missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One EASYseal™ Adhesive Sealer
Protects against evaporation and contamination. Greiner Bio-One™ VIEWseal™ Adhesive Sealer is a standard adhesive foil for seal microplates.
Brand: Greiner Bio-One 676001
Additional Details : Weight : 0.23400kg
Description
- Coated with an acrylate adhesive
- Coverage/storage of microplates (e.g in ELISA applications)
- Protection against evaporation and contamination
- Suitable for optical measurements
- For single use only
Specifications
Transparent | |
Non-sterile | |
100 | |
79.38 mm | |
Acrylic | |
Microplate |
Polyester Film, Acrylic Adhesive | |
135.1 mm | |
135.1 x 79.38 mm | |
Sealer | |
50.8 μm |