Learn More
STEMCELL Technologies EasySep™ Red Blood Cell Lysis Buffer
Red blood cell lysis buffer, 10X concentrate
£70.20 - £494.10
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17108021
|
STEMCELL Technologies
20110 |
10 mL |
£70.20
10mL |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17138021
|
STEMCELL Technologies
20120 |
100 mL |
£494.10
100mL |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood