Learn More
Thermo Scientific™ ABgene™ EasyStrip™ Plus Tube Strip with Attached Flat Caps
The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Flat Caps is designed to reduce the risk of cross-contamination in PCR assays.
Brand: Thermo Scientific™ AB2000
263.21 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Flat Caps is designed to reduce the risk of cross-contamination in PCR assays. The tube strip is compatible with 0.2mL thermal cycler blocks and features individually attached flat caps for quick and easy access to individual samples. In addition, graduated 20μL markers on each tube enable visual volume verification and measurement of reagent and sample additions.For tube strips with ultra clear caps, see Cat. No. AB-2005.
Features:
- Avoid cross contamination with the individually attached caps
- Verify tube reaction volumes with graduated 20μL markings on each tube
- Track samples using the end tabs to label each strip
- Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)
- PCR
Certifications
Certified human DNA, RNase and DNase free
Specifications
Certified human DNA, RNase and DNase free | |
Room Temperature | |
Thermal Cyclers |
Clear | |
250 Strips | |
Flat |