missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Annexin V Apoptosis Detection Kits

Kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane.

Brand:  Invitrogen™ 88-8102-72

 View more versions of this product

Product Code. 14819780

  • £119.00 / 50 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes:
  • 10X Binding Buffer
  • Annexin V PE : 5μL/test, 50 tests Protect from light.
  • 7-AAD Viability Staining Solution : 5μL/test, 100 tests Protect from light.
  • Cat. No. 88-8102-74:
  • 10X Binding Buffer
  • Annexin V PE : 5μL/test, 200 tests Protect from light.
  • 7-AAD Viability Staining Solution : 5μL/test, 2x100 tests Protect from light.
This item is not returnable. View return policy

Description

Description

Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.

Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).

Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

eBioscience Annexin V Apoptosis Detection Kit PE
Contains:
• Annexin V Binding Buffer (10X), 1 bottle (30 mL)
• Annexin V PE, 1 vial (0.25 mL)
• 7-AAD Viability Staining Solution, 1 vial (0.5 mL)

Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits.
Handling Conditions:
Store at 2-8°C. Avoid temperature fluctuations.
PE, 7-AAD
50 Tests
Apoptosis Detection Kit
Wet Ice
Fluorescence Microscope, Flow Cytometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers