Learn More
Fisherbrand™ Ebony Treatment Dish
Protects DNA from damage. Fisherbrand™ Ebony Treatment Dishes are manufactured with a darkened acrylic to reduce exposure to light that may be harmful to DNA.
Brand:  Fisherbrand™ STAINDISH4X
 View more versions of this product
Product Code. 15599901
  • £249.00 / Pack of 4
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from Ebony Acrylic
  • Minimizes exposure to light and reduces background DNA damage
Specifications
Specifications
Ebony
Comet Assay Tank
Acrylic
4