missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ EBSS, no calcium, no magnesium, no phenol red
GREENER_CHOICE

Product Code. 11570466
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11570466

Brand: Gibco™ 14155048

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

EBSS, no calcium, no magnesium, no phenol red

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Gibco™ EBSS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ EBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ EBSS product made in our Grand Island facility (14155-063). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15 to 30°C
Description Earle's Balanced Salt Solution, 500mL, 1 X Concentration, Liquid, 7.0 to 8.1 pH, 36 Months Shelf Life, 270 to 310mOsm/kg Osmolality, With Glucose, Sodium Bicarbonate, Used for Mammalian Cell Culture Applications
Concentration 1 X
Form Liquid
Product Type Earle's Balanced Salt Solution
With Additives Glucose, Sodium Bicarbonate
Without Additives No Calcium, No Magnesium, No Phenol Red
For Use With (Application) Mammalian Cell Culture
pH 7.0 to 8.1
Quantity 500 mL
Regulatory Status For in-vitro diagnostic use
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.