Learn More
Trajan™ EC GC Septa
EC GC septa is designed for long lifetime and low injection port adhesion
Brand: Trajan™ 041842
This item is not returnable.
View return policy
Description
- Combines long injection lifetime, low bleed and low injection port adhesion
- Reliable seal against carrier gas pressure in the inlet
- No bleed or contamination into the GC system
- Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
Long Life EC Septum | |
350°C | |
50/Pk. |
7 mm | |
High Temperature Silicon |