Learn More
Trajan™ EC GC Septa
EC GC septa is designed for long lifetime and low injection port adhesion
Brand:  Trajan™ 041845
 View more versions of this product
Product Code. 10442381
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Combines long injection lifetime, low bleed and low injection port adhesion
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
Specifications
Long Life EC Septum
350°C
50/Pk.
10 mm
High Temperature Silicon