missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ EC GC Septa

EC GC septa is designed for long lifetime and low injection port adhesion

Brand:  Trajan™ 041900

 View more versions of this product

Product Code. 12935731

  • £50.65 / Pack of 25
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Combines long injection lifetime, low bleed and low injection port adhesion
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications

Specifications

Long Life EC Septum
350°C
High Temperature Silicone
9 mm
Varian/Bruker 1177 Injector
25/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.