Learn More
Terumo™ Concentric Luer-Lock Three-Part Syringe
Terumo™ Eccentric Luer-Lock Three-Part Syringes are easy to use with their smooth plunger action and have clear barrels to allow quick recognition of air bubbles for more accurate dosing. With easy to read markings, you can be confident you are delivering the intended dose every time.
Brand:  Terumo™ 1SS02LE1
Product Code. 11793313
  • £12.45 / Pack of 100
Edge
Explore more special offers
Description
Description
  • Silicone coated barrel for a smooth plunger action
  • Clear barrel to allow quick recognition of air bubbles
  • Easy to read markings
  • Latex free
  • Sterile and packed in individually wrapped tamper peel packs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Screen printed graduation
Yes
Single-use
2.5 mL
Sterile
Polypropylene
No Needle Included
Concentric Luer-lock 3-part Syringe
100/Pk.
Concentric Luer Lock
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Terumo™ Concentric Luer-Lock Three-Part Syringe > Volume: 2.5 mL