Learn More
ECD Mouse anti-Human, Unconjugated, Clone: 1D5E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67179-1-IG-20UL
This item is not returnable.
View return policy
Description
The ECD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ECD. This antibody recognizes human, mouse, rat antigen. The ECD antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ECD | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1D5E8 | |
Unconjugated | |
20 μL | |
11319 | |
IgG1 |