missing translation for 'onlineSavingsMsg'
Learn More

ECL Plex Goat anti-Mouse IgG-Cy™ 5 Secondary Antibody, Cytiva

Brand:  Life Science products Cytiva PA45009

Product Code. 11525075

  • £288.00 / 150µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Ms

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG
Cyanine5
150 μg
Purified
Monoclonal
Goat
Secondary
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers