missing translation for 'onlineSavingsMsg'
Learn More

ECL Plex Goat anti-Rabbit IgG-Cy™ 3 Secondary Antibody, Cytiva

Brand:  Life Science products Cytiva 28-9011-06

 View more versions of this product

Product Code. 11580634

  • £386.00 / 150µg
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Rb

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG
Cyanine3
150 μg
Purified
Monoclonal
Goat
Secondary
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ECL Plex Goat anti-Rabbit IgG-Cy™ 3 Secondary Antibody, Cytiva > 150μg; Cy3

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.