missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECL Plex Goat anti-Rabbit IgG-Cy™ 3 Secondary Antibody, Cytiva
Brand: Life Science products Cytiva 28-9011-07
This item is not returnable.
View return policy
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Rb
Western Blotting
Specifications
IgG | |
Cyanine3 | |
600 μg | |
Purified |
Monoclonal | |
Goat | |
Secondary | |
IgG |