missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECL Plex Goat anti-Rabbit IgG-Cy™ 5 Secondary Antibody, Cytiva
Brand: Life Science products Cytiva PA45011
143.45 GBP valid until 2025-12-31
Use promo code "24280" to get your promotional price.
This item is not returnable.
View return policy
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Rb
Western Blotting
Specifications
IgG | |
Cyanine5 | |
150 μg | |
Purified |
Monoclonal | |
Goat | |
Secondary | |
IgG |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ECL Plex Goat anti-Rabbit IgG-Cy™ 5 Secondary Antibody, Cytiva > 150μg; Cy5
Spot an opportunity for improvement?Share a Content Correction