missing translation for 'onlineSavingsMsg'
Learn More

ECL Plex Goat anti-Rabbit IgG-Cy™ 5 Secondary Antibody, Cytiva

Brand:  Life Science products Cytiva PA45012

 View more versions of this product

Product Code. 11555075

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Rb

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG
Cyanine5
600 μg
Purified
Monoclonal
Goat
Secondary
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers