Learn More
Cytiva ECL Select Western Blotting Detection Reagent
Highly intense signal output for detection of medium to very low protein levels. Cytiva ECL Select Western Blotting Detection Reagent is a high sensitivity reagent for chemiluminescent Western blotting detection. Includes Luminol and Peroxide solutions, sufficient for 1000cm2 membrane.
Brand: Cytiva RPN2235SK
Code : GH
Additional Details : Weight : 0.18000kg
Description
High signal intensity makes ECL Select suitable for the most demanding Western blotting applications, allowing for detection of minute protein quantities. Its long lasting signals allow for multiple exposures and a convenient time window between experiment and analysis. Signals can be detected with CCD camera or X-ray film.
- Improved sensitivity: Produces a very high signal intensity
- Quantitative measurements: Linear dynamic range allowing for quantitation of medium to very low abundant proteins
- Works with a wide range of antibody dilutions, reducing the need for lengthy optimization
- Optimized for imaging with ImageQuant LAS 4000 and compatible with Amersham™ Hyperfilm
- Compatible with Rainbow™ Molecular Weight Markers and ECL DualVue Western Blotting Markers
- Optimized for use with Amersham Hybond-P (PVDF) membranes and compatible with Hybond ECL (nitrocellulose) membranes
Specifications
50 mL luminol, 50 mL peroxide | |
Western Blotting Detection Reagent | |
Western blotting detection of medium to very low protein levels |
Safety and Handling
- ECL Select
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360D-May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : imidazole, trometamol, ethanediol
- UNKNOWN ACUTE-4 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 4% of components with unknown hazards to the aquatic environment.