Learn More
BD Eclipse™ Hypodermic Injection Needle with SmartSlip™ Technology
Ensures protection from needlestick injuries after injection. The BD Eclipse™ Hypodermic Injection Needle features the same safety mechanism as the proven BD Eclipse™ Blood Collection Needle, with SmartSlip™ Technology to ensure a secure connection to Luer slip syringes.
Brand:  BD 305895
 View more versions of this product
Product Code. 10891581
  • £381.00 / Pack of 1200
Explore more special offers
Description
Description
  • Unique SmartSlip technology helps to ensure a secure connection to Luer slip syringes
  • Bevel oriented to safety cover accommodates low angle injections
  • One technology for skin injection and blood collection
  • Requires minimal change in injection technique
  • The hub incorporates a plastic clip to encourage user to attach needle more securely to Luer slip syringe
  • Wide textured finger pad provides safe activation area for thumb or forefinger
  • Integral safety cover and extended sidewall to enhance protection
  • Needle remains locked inside activated needle cover
  • Activates with a touch of a finger – no hard surface required for activation
  • Colour coded hub for easy needle size identification