missing translation for 'onlineSavingsMsg'
Learn More

BTX™ ECM 2001 LITE Hybridoma Production System

ECM 2001 LITE Hybridoma Production System is a reliable, affordable and easy-to-use cell fusion and mammalian transfection system.

Brand:  BTX™ 452068

Product Code. 17304990

  • £28400.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features and Benefits:

  • Quick Start Electrofusion menu with 3-step AC-DC-AC protocol structure for speedy access to the most used experimental parameters
  • Storage of up to 1,000 User-Defined protocols with 3-step AC-DC-AC protocol structure
  • Flexibility to inactivate the Electrofusion protocol AC steps for use in square wave electroporation applications, or to inactivate the Electrofusion protocol DC step for embryo manipulation applications
Specifications

Specifications

Hybridoma Production
Color Touchscreen
Non-sterile
5 to 500 V (in 1 V Steps)
Touch Screen Display, Footswitch
100/240 V
ECM 2001 LITE Generator, 2 mL Coaxial Optimization Chamber, 9 mL Coaxial Production Chamber, Female/Female Adapter Set, BTX Cytofusion Medium C 500 mL
505 to 3000 V (in 1 V Steps)
Output Cable
AC Sine Wave, DC Square Wave
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.