Learn More
Sharpsguard™ Eco 0.5 Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco yellow web containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic
Brand:  Sharpsguard™ DD442YLR
Product Code. 17734844
  • £11.05 / Each
Edge
Explore more special offers
Description
Description
  • Sharpsguard™ eco yellow 0.5 with web opening, is ideal for community healthcare workers and easily fitted into a bag or glove compartment.
  • This container is supplied with lid assembled.
  • Apart from a difference in shade of color and an added 'R' on the end of the product code, there is no difference to the safe and easy to use range that you know and trust.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-01-09, 18-02-01, 18-02-02, 18-02-08.
Specifications
Specifications
0.4 L
79 x 45 x 206 mm
1 Ea.
Yellow
26 x 30 mm
88 g