Learn More
Sharpsguard™ Eco 1 Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco 1 container, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic.
Brand:  Sharpsguard™ DD477YLR
 View more versions of this product
Product Code. 17784854
  • £12.50 / Each
Edge
Explore more special offers
Description
Description
  • Sharpsguard™ eco 1 is ideal for phlebotomists. Its push-down, one-piece lid incorporates a vacutainer remover, and will fit securely into our POUDS™ tray range and other carrying trays.
  • This container is already supplied with the lid assembled. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
Specifications
0.84 L
204 x 68 x 119 mm
1 Ea.
Yellow
70 x 32 mm
153 g