missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sharpsguard™ Eco 11.5 Waste Container
The new Sharpsguard™ eco containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
£18.00
Specifications
Capacity (Metric) | 9.27 L |
---|---|
Diameter (Metric) Outer | 275 mm |
Height (Metric) | 249 mm |
Quantity | 1 Ea. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17754854
|
Sharpsguard™
DD476OLR |
Orange |
£18.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17764854
|
Sharpsguard™
DD476YLR |
Yellow |
£18.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sharpsguard™ eco 11.5 is a multi-purpose container featuring a robust pail handle for easy transportation. A wider aperture allows easy insertion of larger items.
- This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
- It is designed to fit safely and securely into our POUDS™ mobile holders and bracket range.
- This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-0
Specifications
9.27 L | |
275 mm | |
249 mm | |
1 Ea. |