Learn More
Sharpsguard™ Eco 2.5 Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
Brand:  Sharpsguard™ DD472OLR
 View more versions of this product
Product Code. 17764844
  • £13.70 / Each
Edge
Explore more special offers
Description
Description
  • Sharpsguard™ eco 2.5 is an ideal general-purpose sharps container, it provides maximum capacity in the minimum working space.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • It is designed to fit safely and securely into our POUDS™ tray range.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
Specifications
2.21 L
Orange
1 Ea.
176 mm
166 mm
232 g