missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sharpsguard™ Eco 22 Waste Container

Click to view available options
Color:
Yellow
Unit Size:
Each
Description
- Sharpsguard™ eco 22 is ideal for theatres, laboratories and sluice rooms with high volumes of hazardous waste. The wide aperture allows economical and convenient disposal of large items.
- This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
- It is designed to fit safely and securely into our POUDS™ mobile holders and bracket range.
- This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
Specifications
| Capacity (Metric) | 17.86 L |
| Diameter (Metric) Outer | 327 mm |
| Color | Yellow |
| Height (Metric) | 343 mm |
| Quantity | 1 Ea. |
| Width (Metric) | 943 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction