missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 65 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE15550
This item is not returnable.
View return policy
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
2 x 6 | |
Aluminum | |
Upright Freezers | |
65 mm |
Microtiterplates | |
12 | |
540 x 135 x 133 mm |