missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Eco Isolation Gowns with Back Laces

Product Code. 17790159
Click to view available options
Color:
Green
Sky Blue
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

These isolation gowns are complete with elasticated wrists and easy tie closures for busy healthcare workers.

  • Light and breathable
  • Collarless model with elasticated cuffs
  • Dorsal closure with laces at waistband and collar
  • Worn by healthcare workers and visitors over clothing
  • Protects patients from risk of external infection

Specifications

Size Universal
For Use With (Application) Dental, Medical
Gender Unisex
No. of Pockets None
Product Type Isolation Gown
Material Polypropylene
Color Yellow
Closure Type Back Ties
Disposable Single-use
Sterility Non-sterile
Weight (Metric) 17 g/m2
Autoclavable Not Autoclavable
No. per Pack 10 Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.