Learn More
Fisherbrand™ ECO 25 Thermoreactor for COD Analysis
Enables the user to determine parameters such as COD, nitrogenous and phosphorated compounds. Fisherbrand™ ECO 25 Thermoreactor for COD Analysis enable the preparing of samples by way of wet digestion for determining metallic and non-metallic elements in organic and inorganic materials.
Brand:  Fisherbrand™ F101A0125
Product Code. 11751304
  • £658.00 / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • COD analyzed in just 30 minutes
  • Acoustic signal and automatic switch-off at the end of each cycle
  • Five working selectable temperatures: (70°, 100°, 120°, 150° and 160°C)
  • Test tube cover, heating block protection and safety thermostat to enhance safety and reliability
Specifications
Specifications
Thermoreactor
70°, 100°, 120°, 150° and 160°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ ECO 25 Thermoreactor for COD Analysis > Product Type: Thermoreactor