Learn More
Thermo Scientific™ Eco32I (EcoRV) (10 U/μL)
The Eco32I (EcoRV) restriction enzyme recognizes GAT^ATC sites and cuts best at 37°C in R buffer (Isoschizomers: EcoRV).
£49.07 - £173.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10369529
|
Thermo Scientific™
ER0301 |
2000 U |
£49.07
2000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11873823
|
Thermo Scientific™
ER0302 |
5 x 2,000 units |
£173.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10829180
|
Thermo Scientific™
ER0305 |
4,000 units |
£78.75
4000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
- Superior quality – stringent quality control and industry leading manufacturing process
- Convenient color-coded Five Buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities
Specifications
10 U/μL |