missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velp Scientifica™ ECO 8 Thermoreactors for COD Analysis
Velp Scientifica™ ECO 8 Thermoreactors for COD Analysis enable the preparing of samples for determining metallic and non-metallic elements in organic and inorganic materials such as minerals, alloys, animal feeds, soils, sediments and organic tissues.
Brand: Velp Scientifica™ F101A0127
This item is not returnable.
View return policy
Description
- Temperature runs up to 160°C
- COD analyzed in just 30 minutes
- Five working temperatures selectable
- Test tube cover, heating block protection and safety thermostat to enhance safety and reliability
- Can process 8 samples in Ø 16 mm test tubes plus one sample in Ø 22 mm
Specifications
COD analysis | |
Temperature settings of 70°, 100°, 120°, 150° and 160°C | |
135 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction