missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolite(+) LC Fluid, MP Biomedicals
Liquid Scintillation Cocktails; Ecolite(+); MP Biomedical; Biodegradable
Brand: MP Biomedicals 0188247501
310.87 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Liquid Scintillation Cocktails | |
1 gal. |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ecolite
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : phenylxylylethane, nonylphenol
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ecolite(+) LC Fluid, MP Biomedicals > 1 gal
Spot an opportunity for improvement?Share a Content Correction