Learn More
Ansell™ Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves Product Code.: 12667011
In Stock
to see stock.
Add to basket
Ansell™ Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves
Product Code. 12667011 Shop All Ansell Products
7
Click to view available options
Size:
7
8
9
10
Product Code. 12667011
Brand: Ansell™ 87190070BP
In Stock
to see stock.
Add to basket
Add to basket
Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves - Extra thin glove for excellent sensitivity. Specially treated to reduce the risk of allergic reactions.
  • Specially treated to reduce the risk of allergic reactions.
  • Thickness of 0.35 mm, provides outstanding sensitivity.
  • Excellent price/quality ratio.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Rubber Latex
Cuff Material Natural Rubber Latex
Coating Material Natural Rubber Latex
Latex Free No
Product Type Extra Thin Gloves
Size 7
Style 87-190
Thickness (Metric) 0.35 mm
Glove Cuff Color Yellow
Glove Cuff Style Straight
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Liquid & mechanical resistant
Series 87-190
Product Title
Ansell™ Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves >